Learn More
PGRP-1b Monoclonal Antibody (186C426), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191734
Description
MA1-91734 detects PGRP1B from human samples. MA1-91734 has been successfully used in immunocytochemistry, Western blot applications. The MA1-91734 immunogen is: Synthetic peptide: CSQRLRELQAHHVHNN conjugated to KLH, corresponding to amino acids 95-110 of Human PGRP1B. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
| PGRP-1b | |
| Monoclonal | |
| 0.5 mg/mL | |
| PGLYRP4 | |
| PGLYRP4 | |
| synthetic peptide CSQRLRELQAHHVHNN conjugated to KLH, corresponding to amino acids 95-110 of Human PGRP1B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 186C426 | |
| Unconjugated | |
| Q96LB8, Q0VB07 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100689249, 310611, 384997, 57115 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.