missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGRMC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110103
This item is not returnable.
View return policy
Description
Immunogen sequence: MAAGDGDVKL GTLGSGSESS NDGGSESPGD AGAAAEGGGW AAAALALLTG.
PGRMC2 (progesterone receptor membrane component 2), also known as DG6 (steroid receptor protein DG6) or PMBP (progesterone membrane-binding protein), is a single pass membrane protein belonging to the cytochrome b5 family (MAPR (membrane associated progesterone receptor) subfamily). Expressed in sperm, PGRMC2 is believed to function as a steroid receptor and may participate in the progesterone-dependent sperm acrosome reaction. PGRMC2 shares approximately 50% overall sequence identity with its close relative PGRMC1. The loss of the gene encoding PGRMC2 is associated with metastasis in uterine endocervical adenocarcinomas, implicating a potential role of PGRMC2 in the suppression of metastasis of endocervical adenocarcinomas.
Specifications
| PGRMC2 | |
| Polyclonal | |
| Unconjugated | |
| Pgrmc2 | |
| 4631434O19Rik; 5730409G06Rik; DG6; Membrane-associated progesterone receptor component 2; Pgrmc2; PMBP; progesterone membrane binding protein; Progesterone membrane-binding protein; progesterone receptor membrane component 2; steroid receptor protein DG6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10424, 70804 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.89 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O15173, Q80UU9 | |
| Pgrmc2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-50 of human PGRMC2 (NP_0063113). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction