missing translation for 'onlineSavingsMsg'
Learn More

PGRMC2 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18429442 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429442 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429442 Supplier Novus Biologicals Supplier No. NBP149716R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PGRMC2 Polyclonal antibody specifically detects PGRMC2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGRMC2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DG6progesterone membrane binding protein, membrane-associated progesterone receptor component 2, PMBPProgesterone membrane-binding protein, progesterone receptor membrane component 2, Steroid receptor protein DG6
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Progesterone Receptor Membrane Component 2 using the numbering given in entry NP_006311.1 (GeneID 10424).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10424
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.