missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGRMC2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49716AF700
This item is not returnable.
View return policy
Description
PGRMC2 Polyclonal antibody specifically detects PGRMC2 in Human samples. It is validated for Western Blot
Specifications
| PGRMC2 | |
| Polyclonal | |
| Western Blot | |
| DG6progesterone membrane binding protein, membrane-associated progesterone receptor component 2, PMBPProgesterone membrane-binding protein, progesterone receptor membrane component 2, Steroid receptor protein DG6 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Progesterone Receptor Membrane Component 2 using the numbering given in entry NP_006311.1 (GeneID 10424). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 10424 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction