missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGRMC1 Polyclonal Antibody

PGRMC1 Polyclonal Antibody
Brand: Invitrogen PA542919
This item is not returnable.
View return policy
Description
PGRMC1 Polyclonal Antibody for Western Blot, IHC
Specifications
| PGRMC1 | |
| Polyclonal | |
| Unconjugated | |
| PGRMC1 | |
| 25dx; 25-Dx; AA415812; Acidic 25 kDa protein; Dap1; HGNC:16090; HPR6.6; IZA; Lewi; membrane steroid binding protein; membrane-associated progesterone receptor component 1; membrane-bound progesterone receptor; MPR; Pgrmc; Pgrmc1; PPMR; progesterone binding protein; progesterone receptor membrane component 1; steroid membrane binding protein; VEMA; Ventral midline antigen | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00264 | |
| PGRMC1 | |
| Synthetic peptide directed towards the N-terminal of human PGRMC1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction