missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGR1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | PGR1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PGR1 Polyclonal antibody specifically detects PGR1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| PGR1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 22949 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 15-oxoprostaglandin 13-reductase, EC 1.3.1.-, EC 1.3.1.48, EC 1.3.1.74, FLJ99229, leukotriene B4 12-hydroxydehydrogenase, LTB4DH, MGC34943, NADP-dependent leukotriene B4 12-hydroxydehydrogenase, PGR1, PRG-1, prostaglandin reductase 1, ZADH3, zinc binding alcohol dehydrogenase domain containing 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-115 of human PTGR1 (NP_036344.2). MVRTKTWTLKKHFVGYPTNSDFELKTAELPPLKNGEVLLEALFLTVDPYMRVAAKRLKEGDTMMGQQVAKVVESKNVALPKGTIVLASPGWTTHSISDGKDLEKLLTEWPDTIPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title