missing translation for 'onlineSavingsMsg'
Learn More

PGPEP-1 Antibody [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30764123 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30764123 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30764123 Supplier Novus Biologicals Supplier No. AF6278MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

PGPEP-1 Polyclonal antibody specifically detects PGPEP-1 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGPEP-1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.19.3, Pcp, PGPIFLJ20208, PGP-IPAP-I, PGPMGC10812, Pyroglutamyl aminopeptidase I, pyroglutamyl-peptidase 1, pyroglutamyl-peptidase I5-oxoprolyl-peptidase, Pyrrolidone-carboxylate peptidase
Host Species Sheep
Immunogen E. coli-derived recombinant human PGPEP-1, Met1-His209, Accession # Q9NXJ5
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 54858
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.