missing translation for 'onlineSavingsMsg'
Learn More

PGMI-004A, MedChemExpress

Product Code. 16484536
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484536 100 mg 100mg
16466007 5 mg 5mg
16439826 50 mg 50mg
16433288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484536 Supplier Medchem Express Supplier No. HY101143
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PGMI-004A is a potent phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 13.1 μM.

PGMI-004A is a potent phosphoglycerate mutase 1 (PGAM1) inhibitor with an IC50 of 13.1 μM.

Specifications

Chemical Name or Material PGMI-004A
CAS 1313738-90-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H12F3NO6S
Quantity 100 mg
Solubility Information DMSO : ≥ 125 mg/mL (269.76 mM)
SMILES O=S(C(C=C1C2=O)=C(O)C(O)=C1C(C3=C2C=CC=C3)=O)(NC4=CC=C(C(F)(F)F)C=C4)=O
Molecular Weight (g/mol) 463.38
Formula Weight 463.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.