missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGM5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548880
This item is not returnable.
View return policy
Description
PGM5 Polyclonal Antibody for Western Blot
Component of adherens-type cell-cell and cell-matrix junctions. Lacks phosphoglucomutase activity.
Specifications
| PGM5 | |
| Polyclonal | |
| Unconjugated | |
| PGM5 | |
| Aciculin; PGM5; PGMRP; PGM-RP; phosphoglucomutase 5; phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 5239 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q15124 | |
| PGM5 | |
| KLH conjugated synthetic peptide between 404-430 amino acids from the C-terminal region of human PGM5. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction