missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556249
This item is not returnable.
View return policy
Description
Immunogen sequence: KPPQGMEIKS NERCCSFDGD ADRIVYYYHD ADGHFHLIDG DKIATLISSF LKELLVEIGE SLNIGVVQTA YANGSSTRYL EEVMKVPVYC TKTGV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 89%.
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.
Specifications
| PGM3 | |
| Polyclonal | |
| Unconjugated | |
| Pgm3 | |
| 2810473H05Rik; acetylglucosamine phosphomutase; Agm1; BB187688; C77933; GlcNAc-P mutase; IMD23; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Pgm3; Pgm-3; Phosphoacetylglucosamine mutase; phosphoglucomutase 3; phosphoglucomutase-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5238 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95394 | |
| Pgm3 | |
| Recombinant protein corresponding to Human PGM3. Recombinant protein control fragment (Product #RP-94592). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction