Learn More
Invitrogen™ PGM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114087
Description
Immunogen sequence: GVVQTAYANG SSTRYLEEVM KVPVYCTKTG VKHLHHKAQE FDIGVYFEAN For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 93%.
Catalyzes the conversion of GlcNAc-6-P into GlcNAc-1-P during the synthesis of uridine diphosphate/UDP-GlcNAc, a sugar nucleotide critical to multiple glycosylation pathways including protein N- and O-glycosylation.
Specifications
| PGM3 | |
| Polyclonal | |
| Unconjugated | |
| Pgm3 | |
| 2810473H05Rik; acetylglucosamine phosphomutase; Agm1; BB187688; C77933; GlcNAc-P mutase; IMD23; N-acetylglucosamine-phosphate mutase; N-acetylglucosamine-phosphate mutase 1; PAGM; PGM 3; Pgm3; Pgm-3; Phosphoacetylglucosamine mutase; phosphoglucomutase 3; phosphoglucomutase-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5238 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95394 | |
| Pgm3 | |
| Synthetic peptide directed towards the middle region of human PGM3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.