missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM3 Antibody (OTI5C6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73364B
This item is not returnable.
View return policy
Description
PGM3 Monoclonal specifically detects PGM3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| PGM3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PGM3 | |
| Full length recombinant protein of human PGM3 (NP_056414) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI5C6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| AGM1PAGMAcetylglucosamine phosphomutase, DKFZP434B187, EC 5.4.2, EC 5.4.2.3, FLJ11614, FLJ13623, N-acetylglucosamine-phosphate mutase, N-acetylglucosamine-phosphate mutase 1, PGM 3, phosphoacetylglucosamine mutase, phosphoglucomutase 3, Phosphoglucomutase-3 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 5238.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction