missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PGM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29668
This item is not returnable.
View return policy
Description
PGM2 Polyclonal antibody specifically detects PGM2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PGM2 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| EC 5.4.2, EC 5.4.2.2, EC 5.4.2.7, FLJ10983, Glucose phosphomutase 2, PGM 2, Phosphodeoxyribomutase, phosphoglucomutase 2, phosphoglucomutase-2, Phosphopentomutase | |
| Between 175 and 225 | |
| 100 μg | |
| metabolism | |
| 55276 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction