missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35558-100ul
This item is not returnable.
View return policy
Description
PGM2 Polyclonal antibody specifically detects PGM2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PGM2 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| EC 5.4.2, EC 5.4.2.2, EC 5.4.2.7, FLJ10983, Glucose phosphomutase 2, PGM 2, Phosphodeoxyribomutase, phosphoglucomutase 2, phosphoglucomutase-2, Phosphopentomutase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PGM2 (NP_060760.2).,, Sequence:, MAAPEGSGLGEDARLDQETAQWLRWDKNSLTLEAVKRLIAEGNKEELRKCFGARMEFGTAGLRAAMGPGISRMNDLTIIQTTQGFCRYLEKQFSDLKQKGIVISFDARAHPSSGGSSRRFARLAATTFIS | |
| 100 μL | |
| metabolism | |
| 55276 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction