missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19787
This item is not returnable.
View return policy
Description
PGM2 Polyclonal specifically detects PGM2 in Human samples. It is validated for Western Blot.
Specifications
| PGM2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| EC 5.4.2, EC 5.4.2.2, EC 5.4.2.7, FLJ10983, Glucose phosphomutase 2, PGM 2, Phosphodeoxyribomutase, phosphoglucomutase 2, phosphoglucomutase-2, Phosphopentomutase | |
| Rabbit | |
| 68 kDa | |
| 0.1 mL | |
| metabolism | |
| 55276 | |
| Human | |
| IgG |
| Western Blot | |
| 0.41mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q96G03 | |
| PGM2 | |
| Recombinant protein encompassing a sequence within the center region of human PGM2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Chicken (83%), Xenopus tropicalis (80%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction