Learn More
Invitrogen™ PGM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535947
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Rat (98%), Zebrafish (82%), Xenopus laevis (92%), Pig (96%), Rabbit (98%), Chicken (93%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphoglucomutases (PGM; EC 5.4.2.2) catalyze the transfer of phosphate between the 1 and 6 positions of glucose. Isozymes of PGM are monomeric, with molecular masses of about 60 kD, and are encoded by several genes, including PGM1. In most cell types, PGM1 isozymes predominate, representing about 90% of total PGM activity. One exception is red cells, where PGM2 (MIM 172000) is a major isozyme (Putt et al., 1993 [PubMed 8257433]). [supplied by OMIM].
Specifications
| PGM1 | |
| Polyclonal | |
| Unconjugated | |
| PGM1 | |
| 3230402E02Rik; Aciculin; CDG1T; Glucose phosphomutase 1; glucose phosphomutase 2; GSD14; PGM; PGM 1; PGM 2; Pgm1; Pgm-1; Pgm2; PGM5; PGMRP; PGM-RP; Phosphodeoxyribomutase; phosphoglucomutase 1; phosphoglucomutase 2; phosphoglucomutase 5; phosphoglucomutase isoform 2; phosphoglucomutase isoform1; phosphoglucomutase-1; Phosphoglucomutase-2; phosphoglucomutase-like protein 5; Phosphoglucomutase-related protein; phosphopentomutase; zgc:63718 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5236 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.82 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P36871 | |
| PGM1 | |
| Recombinant protein encompassing a sequence within the center region of human PGM1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.