missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM1 Antibody (CL3301) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59026G
This item is not returnable.
View return policy
Description
PGM1 Monoclonal antibody specifically detects PGM1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PGM1 | |
| Monoclonal | |
| DyLight 488 | |
| EC 5.4.2, EC 5.4.2.2, Glucose phosphomutase 1, GSD14, PGM 1, phosphoglucomutase 1, phosphoglucomutase-1 | |
| This antibody was developed using a recombinant protein derived from P36871, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5236 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3301 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction