missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23029-1-AP-150UL
This item is not returnable.
View return policy
Description
Peptidoglycan recognition proteins, such as PGRPL, are part of the innate immune system and recognize peptidoglycan, a ubiquitous component of bacterial cell walls.Specifications
| PGLYRP2 | |
| Polyclonal | |
| Unconjugated | |
| PGLYRP2 | |
| HMFT0141, PGLYRP2, PGLYRPL, PGRP L, PGRPL, tagL, tagL alpha, tagl beta, TAGL like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114770 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96PD5 | |
| PGLYRP2 | |
| PGLYRP2 Fusion Protein Ag19336 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction