missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP1/PGRP-S Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£356.00 - £559.00
Specifications
| Antigen | PGLYRP1/PGRP-S |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604274
|
Novus Biologicals
NBP3-21331-100ul |
100 μg |
£559.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649694
|
Novus Biologicals
NBP3-21331-25ul |
25 μg |
£356.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGLYRP1/PGRP-S Polyclonal antibody specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for ImmunofluorescenceSpecifications
| PGLYRP1/PGRP-S | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS, pH 7.2, 40% glycerol | |
| 8993 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VPRNEWKALASECAQHLSLPLRYVVVSHTAGSSCNTPASCQQQARNVQHYHMKTLGWCDVGYNF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title