missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP1/PGRP-S Antibody (Novo Nordisk patent anti-PGLYRP1) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28593JF585
This item is not returnable.
View return policy
Description
PGLYRP1/PGRP-S Monoclonal antibody specifically detects PGLYRP1/PGRP-S in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| PGLYRP1/PGRP-S | |
| Monoclonal | |
| Janelia Fluor 585 | |
| peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L | |
| PGLYRP1/ PGRP-S | |
| 0.1 mL | |
| Apoptosis | |
| 8993 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| Novo Nordisk patent anti-PGLYRP1 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction