missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGLS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531678
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, PC-3, U87-MG, SK-N-SH. Predicted reactivity: Mouse (88%), Rat (88%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.
Specifications
| PGLS | |
| Polyclonal | |
| Unconjugated | |
| PGLS | |
| 1110030K05Rik; 6PGL; 6PGL {ECO:0000250; 6-phosphogluconolactonase; 6-phosphogluconolactonase {ECO:0000250; AI447866; epididymis secretory protein Li 304; HEL-S-304; hypothetical protein LOC616120; Pgls; pgls {ECO:0000250; Plgs; UniProtKB:O95336} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25796, 66171 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95336, Q9CQ60 | |
| PGLS | |
| Recombinant protein encompassing a sequence within the center region of human PGLS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction