missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGLS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598053
This item is not returnable.
View return policy
Description
PGLS Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
PGLS belongs to the glucosamine/galactosamine-6-phosphate isomerase family, 6-phosphogluconolactonase subfamily. It is implicated in the hydrolysis of 6-phosphogluconolactone to 6-phosphogluconate.
Specifications
| PGLS | |
| Polyclonal | |
| Unconjugated | |
| PGLS | |
| 1110030K05Rik; 6PGL; 6PGL {ECO:0000250; 6-phosphogluconolactonase; 6-phosphogluconolactonase {ECO:0000250; AI447866; epididymis secretory protein Li 304; HEL-S-304; hypothetical protein LOC616120; Pgls; pgls {ECO:0000250; Plgs; UniProtKB:O95336} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25796, 66171 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.94 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95336, Q9CQ60 | |
| PGLS | |
| Recombinant Human 6-phosphogluconolactonase protein (1-258AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction