missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGL2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | PGL2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18611962
|
Novus Biologicals
NBP2-94039-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693402
|
Novus Biologicals
NBP2-94039-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGL2 Polyclonal antibody specifically detects PGL2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| PGL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 54949 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| C11orf79, FLJ20487, hSDH5, paraganglioma or familial glomus tumors 2, PGL2chromosome 11 open reading frame 79, SDH5SDH assembly factor 2, succinate dehydrogenase assembly factor 2, mitochondrial, succinate dehydrogenase complex assembly factor 2, Succinate dehydrogenase subunit 5, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-166 of human SDHAF2 (NP_060311.1). MAVSTVFSTSSLMLALSRHSLLSPLLSVTSFRRFYRGDSPTDSQKDMIEIPLPPWQERTDESIETKRARLLYESRKRGMLENCILLSLFAKEHLQHMTEKQLNLYDRLINEPSNDWDIYYWATEAKPAPEIFENEVMALLRDFAKNKNKEQRLRAPDLEYLFEKPR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title