missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PGK2 |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679097
|
Novus Biologicals
NBP2-68964-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687298
|
Novus Biologicals
NBP2-68964 |
100 μg |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGK2 Polyclonal antibody specifically detects PGK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PGK2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 5232 | |
| IgG | |
| Protein A purified |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| dJ417L20.2, EC 2.7.2.3, PGK-2, PGKB, PGKPS, phosphoglycerate kinase 2, phosphoglycerate kinase autosomal pseudogene, Phosphoglycerate kinase, testis specific | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GCITIIGGGDTATCCAKWNTEDKVSHVSTGGGASLELLEGKVLP | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title