missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGK1 Rabbit anti-Human, Mouse, Rat, Clone: 4B3Q3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15453-100UL
This item is not returnable.
View return policy
Description
PGK1 Monoclonal antibody specifically detects PGK1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PGK1 | |
| Monoclonal | |
| Unconjugated | |
| Cell migration-inducing gene 10 protein, EC 2.7.2.3, MGC142128, MGC8947, MIG10, PGKAMGC117307, phosphoglycerate kinase 1, Primer recognition protein 2, PRP 2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PGK1 (P00558). | |
| 100 μg | |
| Prostate Cancer | |
| 5230 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4B3Q3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction