missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PGK1 Monoclonal Antibody (1086CT10.2.1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537712

Product Code. 17167298

  • £464.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PGK1 Monoclonal Antibody for Flow, IHC (P), Western Blot

Also known as ATP:3-phosphoglycerate 1-phosphotransferase showed that the plasmin reductase isolated from conditioned medium of fibrosarcoma cells is the glycolytic enzyme phosphoglycerate kinase. They concluded that phosphoglycerate kinase not only functions in glycolysis but is secreted by tumor cells and participates in the angiogenic process as a disulfide reductase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PGK1
Monoclonal
0.25 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P00558, P09411
PGK1
KLH conjugated synthetic peptide between 1-417 amino acids from human.
200 μL
Primary
Human, Mouse
Antibody
IgG2a κ
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1086CT10.2.1
Unconjugated
PGK1
cell migration-inducing gene 10 protein; epididymis secretory sperm binding protein Li 68p; HEL-S-68p; MIG10; OK/SW-cl.110; Pgk; Pgk1; Pgk-1; PGKA; Phosphoglycerate kinase; phosphoglycerate kinase 1; Primer recognition protein 2; PRP 2; YCR012W; YCR12W
Mouse
Protein G, Size-exclusion - Dialysis
RUO
18655, 5230
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.