missing translation for 'onlineSavingsMsg'
Learn More

PGK1 Antibody (2F4), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18421764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421764 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421764 Supplier Novus Biologicals Supplier No. NBP259392AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PGK1 Monoclonal antibody specifically detects PGK1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGK1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2F4
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Cell migration-inducing gene 10 protein, EC 2.7.2.3, MGC142128, MGC8947, MIG10, PGKAMGC117307, phosphoglycerate kinase 1, Primer recognition protein 2, PRP 2
Host Species Mouse
Immunogen Recombinant human PGK1 (1-417aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5230
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.