missing translation for 'onlineSavingsMsg'
Learn More

PGEA1 Antibody (2F2), Novus Biologicals™

Product Code. 18338629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338629 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338629 Supplier Novus Biologicals Supplier No. H00025776M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PGEA1 Monoclonal antibody specifically detects PGEA1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGEA1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2F2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_056188
Gene Alias arb1, ARPP-binding protein, C22orf2chromosome 22 open reading frame 2, Cby, CBYgolgi and endoplasmic reticulum associated 1, chibby homolog 1 (Drosophila), coiled-coil protein PIGEA-14, Cytosolic leucine-rich protein, HS508I15A, PGEA1, PIGEA-14Chibby, PKD2 interactor, Golgi and endoplasmic reticulum-associated 1, polycystin-2 interactor, Golgi- and endoplasmic reticulum-associated protein, protein chibby homolog 1
Host Species Mouse
Immunogen CBY1 (NP_056188, 18 a.a. ~ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SASLSNLHSLDRSTREVELGLEYGSPTMNLAGQSLKFENGQWIAETGVSGGVDRREVQRLRRRNQQLEEENNLLRLKVDILLDMLSESTAESHLMEKELDELRISRKRK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 25776
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.