missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The Non-species specific PGE1 ELISA Kit (Colorimetric) measures non-species specific PGE1 in culture supernatants, plasma, serum, saliva, and urine from any species. cited sample types include simulated interstitial lung fluid.
- Standard Curve Range: 4.88 - 5,000 pg/mL
- Sensitivity: 5.58 pg/mL
- Assay Type: Competitive ELISA
- Inter-Assay: Mean 8.8%
- Intra-Assay: Mean 9.3%
- Spike Recovery:
- Tissue Culture Media - 90-110%
- Human Saliva - 107.2%
- Human Urine - 109.9%
- Human Serum - 87.0%
- Human Plasma - 107.7%
- Sample Volume: 100 μL
- Components:
- Wash buffer concentrate
- Standard
- Stop solution
- pNpp Substrate
- Conjugate
- DxS IgG Microtiter plate
- Antibody
- Assay buffer
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?