missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD2 Synthase/PTGDS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | PGD2 Synthase/PTGDS |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694867
|
Novus Biologicals
NBP3-00115-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618637
|
Novus Biologicals
NBP3-00115-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGD2 Synthase/PTGDS Polyclonal antibody specifically detects PGD2 Synthase/PTGDS in Human samples. It is validated for ELISASpecifications
| PGD2 Synthase/PTGDS | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mast Cell Markers, Neuroscience, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5730 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase | |
| Produced in rabbits immunized with purified, recombinant Human PGD2 Synthase/PTGDS (Accession#: P41222; Met1-Gln190) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title