missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD2 Synthase/PTGDS Antibody (07), FITC , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06151F
This item is not returnable.
View return policy
Description
PGD2 Synthase/PTGDS Monoclonal antibody specifically detects PGD2 Synthase/PTGDS in Human samples. It is validated for ELISA
Specifications
| PGD2 Synthase/PTGDS | |
| Monoclonal | |
| FITC | |
| Beta-trace protein, cerebrin-28, EC 5.3.99.2, Glutathione-independent PGD synthase, glutathione-independent PGD synthetase, lipocalin-type prostaglandin D synthase, Lipocalin-type prostaglandin-D synthase, L-PGDS, PDS, PGD2, PGD2 synthase, PGDS2, PGDSLPGDS, prostaglandin D synthase, prostaglandin D2 synthase (21kD, brain), prostaglandin D2 synthase 21kDa (brain), Prostaglandin-D2 synthase, prostaglandin-H2 D-isomerase | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PGD2 Synthase/PTGDS (P41222; Met1-Gln190). | |
| 0.1 mL | |
| Mast Cell Markers, Neuroscience, Signal Transduction | |
| 5730 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 07 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction