missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD2-IN-1, MedChemExpress
Description
PGD2-IN-1 is an antagonist of DP extracted from patent WO 2006044732 A2, example 15 (d)
Specifications
Specifications
| Chemical Name or Material | PGD2-IN-1 |
| CAS | 885066-67-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H23Cl2N3O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 25 mg/mL (54.31 mM) |
| SMILES | O=C(O)C(C)(C)C1=CC=CC(C2=NC(OC)=NC(NCCC3=CC=C(Cl)C=C3Cl)=C2)=C1 |
| Molecular Weight (g/mol) | 460.35 |
| Formula Weight | 460.35 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?