missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD Rabbit anti-Human, Mouse, Rat, Clone: 9A2M7, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | PGD |
|---|---|
| Clone | 9A2M7 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359085
|
Novus Biologicals
NBP3-15277-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18397335
|
Novus Biologicals
NBP3-15277-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGD Monoclonal antibody specifically detects PGD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| PGD | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5226 | |
| IgG | |
| Affinity purified |
| 9A2M7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 6PGD, EC 1.1.1.44,6-phosphogluconate dehydrogenase, decarboxylating, PGDH, phosphogluconate dehydrogenase | |
| A synthetic peptide corresponding to a sequence within amino acids 384-483 of human PGD (P52209). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title