missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGCP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16601-1-AP-20UL
This item is not returnable.
View return policy
Description
PGCP is a metallopeptidase that belongs to the peptidase M28 family. The encoded protein may catalyze the cleavage of dipeptides with unsubstituted terminals into amino acids.Specifications
| PGCP | |
| Polyclonal | |
| Unconjugated | |
| CPQ | |
| CPQ, LCH1, Lysosomal dipeptidase, PGCP, plasma glutamate carboxypeptidase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10404, 54381 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9WVJ3, Q9Y646 | |
| Cpq | |
| PGCP Fusion Protein Ag9770 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction