missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGCP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547602
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
PGCP is a metallopeptidase that belongs to the peptidase M28 family. The encoded protein may catalyze the cleavage of dipeptides with unsubstituted terminals into amino acids.
Specifications
| PGCP | |
| Polyclonal | |
| Unconjugated | |
| Cpq | |
| 1190003P12Rik; 2610034C17Rik; aminopeptidase; blood plasma glutamate carboxypeptidase; carboxypeptidase Q; Cpq; Hematopoietic lineage switch 2; hematopoietic lineage switch 2 related protein; Hls2; hls2-rp; LAL; Lal-1; LCH1; LDP; liver annexin like-1; liver annexin-like protein 1; Lysosomal dipeptidase; Pgcp; Plasma glutamate carboxypeptidase; Ser-Met dipeptidase | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 54381 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9WVJ3 | |
| Cpq | |
| Mouse myeloma cell line NS0-derived recombinant mouse PGCP Lys19-Ser470. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction