Learn More
PGCF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA131197
Description
PA1-31197 detects PGCF from rat, mouse, human samples. PA1-31197 has been successfully used in Western blot applications. The PA1-31197 immunogen is: Synthetic peptide for particulate guanylyl cyclase F. All peptides were amidated for conjugation. The PGCF selective antibodies were prepared against a unique peptide selective for only PGCF that maps at or close to the C termi
Specifications
| PGCF | |
| Polyclonal | |
| GUCY2F | |
| GUCY2F | |
| Synthetic peptide for particulate guanylyl cyclase F. All peptides were amidated for conjugation. The PGCF selective antibodies were prepared against a unique peptide selective for only PGCF that maps at or close to the C terminal region of the protein. ( | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| P51841, Q5SDA5, P51842 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116556, 245650, 2986 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.