missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPARGC1A Polyclonal Antibody

Product Code. 15967840
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15967840 100 μL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15967840 Supplier Invitrogen™ Supplier No. PA572948
  • ONLINE EXCLUSIVE PRICE
    £451.00
    £356.65 / 100µL
    Save £94.35
    20% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Expected from sequence similarity: Mouse. In IHC-P, staining is very strong in the nucleus with some cytoplasmic staining. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.

PGC1 alpha is a transcriptional coactivator that regulates the genes involved in energy metabolism. It interacts with PPARgamma, which permits it's interaction with multiple transcription factors. PGC-1 Alpha can interact with, and regulate the activities of, cAMP response element binding protein (CREB) and nuclear respiratory factors (NRFs). It provides a direct link between external physiological stimuli and the regulation of mitochondrial biogenesis, and is a major factor that regulates muscle fiber type determination. It may be also involved in controlling blood pressure, regulating cellular cholesterol homoeostasis, and the development of obesity.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen PPARGC1A
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ppargc1a
Gene Accession No. O70343, Q865B6, Q9QYK2, Q9UBK2
Gene Alias A830037N07Rik; ENSMUSG00000079510; Gm11133; I79_015506; LEM6; Ligand effect modulator 6; ligand effect modulator-6; L-PGC-1alpha; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5; peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT; peroxisome proliferator-activated receptor gamma coactivator 1-alpha; peroxisome proliferator-activated receptor gamma coactivator-1 alpha; peroxisome proliferator-activated receptor gamma, coactivator 1 alpha; Pgc1; PGC-1; pgc-1 alpha; PGC-1(alpha); PGC1A; PGC-1alpha; PGC-1-alpha; Pgc-1alphaa; PGC-1v; Pgco1; PPAR gamma coactivator variant form; PPAR Gamma Coactivator-1; PPARG coactivator 1 alpha; PPARgamma coactivator 1alpha; PPAR-gamma coactivator 1-alpha; PPARGC1; PPARGC1A; PPARGC-1-alpha
Gene Symbols Ppargc1a
Host Species Rabbit
Immunogen Recombinant protein made to an internal portion of the human PGC-1 alpha protein (within residues 400-550).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100753651, 100861288, 10891, 19017, 397013, 443270, 479127, 83516
Target Species Human, Mouse, Rat, Canine, Sheep, Hamster, Pig, Rodent, Goat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.