missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGC1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-1750
This item is not returnable.
View return policy
Description
PGC1 alpha Polyclonal specifically detects PGC1 alpha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PGC1 alpha | |
| Polyclonal | |
| Western Blot, Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9UBK2 | |
| PPARGC1A | |
| The immunogen recognized by this PGC1 alpha Antibody maps to a region between residue 250 and 300 of human peroxisome proliferative activated receptor, gamma, coactivator 1, alpha using the numbering given in SwissProt entry Q9UBK2 (Gene ID 10891). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The epitope recognized by PGC1 alpha Antibody maps to a region between residue 250 and 300 of human PGC1 alpha using the numbering given in SwissProt entry Q9UBK2 (Gene ID: 10891). | |
| Store at 4C. Do not freeze. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC-1-alpha | |
| Rabbit | |
| 91 kDa | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, mTOR Pathway, Neuroscience | |
| 10891 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction