missing translation for 'onlineSavingsMsg'
Learn More

PGC1 alpha Antibody (1G8), Novus Biologicals™

Mouse Monoclonal Antibody

£161.00 - £416.00

Specifications

Antigen PGC1 alpha
Clone 1G8
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18637005
View Documents
Novus Biologicals
NBP2-37562-0.025ml
0.025 mL
£161.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18103999
View Documents
Novus Biologicals
NBP2-37562
0.1 mL
£416.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PGC1 alpha Monoclonal specifically detects PGC1 alpha in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PGC1 alpha
Western Blot 1:500 - 1:2000, ELISA 1:10000
Monoclonal
Ascites
RUO
Human
LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC-1-alpha
PPARGC1A
IgG2b
Unpurified
1G8
Western Blot, ELISA
Unconjugated
Mouse
Cancer, Cholesterol Metabolism, Chromatin Research, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Neuroscience, Transcription Factors and Regulators
Ascites with 0.03% Sodium Azide
10891
This PGC1 alpha Antibody (1G8) was developed against a purified recombinant fragment of human PGC-1alpha expressed in E. Coli.
Primary
91 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.