missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556831
This item is not returnable.
View return policy
Description
Immunogen sequence: IQVPNQEFGL SENEPGTNFV YAQFDGIMGL AYPALSVDEA TTAMQGMVQE GALTSPVFSV YLSNQQGSSG GAVVFGGVDS SLYTGQI Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 78%.
Pepsin is one of the main proteolytic enzymes secreted by the gastric mucosa. Pepsin consists of a single polypeptide chain and arises from its precursor, pepsinogen, by removal of a 41 amino acid segment from the N-terminus. Pepsinogen is synthesized in the stomach lining, and hydrochloric acid, also produced by the gastric mucosa, is necessary to convert the inactive enzyme and to maintain the optimum acidity (pH 1-3) for pepsin function. Pepsin is particularly effective in cleaving peptide bonds involving aromatic amino acids. Pepsin shows extremely broad specificity, and although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. The amino acid composition of Pepsin C differs from those of pepsinogen and pepsin especially in the content of basic amino acids, glutamic acid, aspartic acid, leucine and isoleucine.
Specifications
| PGC | |
| Polyclonal | |
| Unconjugated | |
| Pgc | |
| 2210410L06Rik; Gastricsin; pep carboxylase; PEPC; pepcase; pepsin C; pepsinogen C; pepsinogen group II; PG1; Pg-1; Pgc; PGII; preprogastricsin; progastricsin; progastricsin (pepsinogen C); Upg1; Upg-1; Urinary pepsinogen 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5225 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20142 | |
| Pgc | |
| Recombinant protein corresponding to Human PGC. Recombinant protein control fragment (Product #RP-94665). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction