missing translation for 'onlineSavingsMsg'
Learn More

PGC-1 beta Antibody, DyLight 350, Novus Biologicals™

Product Code. 18413138 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413138 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413138 Supplier Novus Biologicals Supplier No. NBP128722UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PGC-1 beta Polyclonal antibody specifically detects PGC-1 beta in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PGC-1 beta
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp686C1790, ERRL1, PERCFLJ14284, peroxisome proliferator-activated receptor gamma, coactivator 1 beta, PGC1, PGC1Bgamma, coactivator 1, beta, PPARGC1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human peroxisome proliferator-activated receptor gamma coactivator 1 beta using the numbering given in entry NP_573570.2 (GeneID 133522).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Diabetes Research, Hypoxia, Lipid and Metabolism, Mitochondrial Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 133522
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.