missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PGC-1 beta Polyclonal antibody specifically detects PGC-1 beta in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PGC-1 beta |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | DKFZp686C1790, ERRL1, PERCFLJ14284, peroxisome proliferator-activated receptor gamma, coactivator 1 beta, PGC1, PGC1Bgamma, coactivator 1, beta, PPARGC1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 850-950 of human PGC1 beta (NP_001166170.1). SSRELKRRFEVFGEIEECEVLTRNRRGEKYGFITYRCSEHAALSLTKGAALRKRNEPSFQLSYGGLRHFCWPRYTDYDSNSEEALPASGKSKYEAMDFDSL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?