missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGC-1 beta Antibody (6C3F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37249
This item is not returnable.
View return policy
Description
PGC-1 beta Monoclonal specifically detects PGC-1 beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| PGC-1 beta | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| DKFZp686C1790, ERRL1, PERCFLJ14284, peroxisome proliferator-activated receptor gamma, coactivator 1 beta, PGC1, PGC1Bgamma, coactivator 1, beta, PPARGC1 | |
| Mouse | |
| 113.2 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Diabetes Research, Hypoxia, Lipid and Metabolism, Mitochondrial Markers, Neuroscience, Signal Transduction | |
| 133522 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 6C3F6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PPARGC1B | |
| Purified recombinant fragment of human PPARGC1B (AA: 195-414) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction