missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGBD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559343
This item is not returnable.
View return policy
Description
Immunogen sequence: VLTLVNDLLG QGYCVFLDNF NISPMLFREL HQNRTDAVGT ARLNRKQIPN DLKKRIAKGT TVARFCGELM ALKWCDGKEV TMLSTFHNDT VIEVNNRNGK KTKR Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 19%.
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families.
Specifications
| PGBD4 | |
| Polyclonal | |
| Unconjugated | |
| PGBD4 | |
| PGBD4; piggyBac transposable element derived 4; piggyBac transposable element-derived protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 161779 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DM1 | |
| PGBD4 | |
| Recombinant protein corresponding to Human PGBD4. Recombinant protein control fragment (Product #RP-98175). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction