missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55392-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to PGAP1.
Involved in inositol deacylation of GPI-anchored proteins. GPI inositol deacylation may important for efficient transport of GPI-anchored proteins from the endoplasmic reticulum to the Golgi.Specifications
| PGAP1 | |
| Polyclonal | |
| Unconjugated | |
| PGAP1 | |
| Bst1, GPI inositol deacylase, hPGAP1, ISPD3024, PGAP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 241062, 80055 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3UUQ7, Q75T13 | |
| PGAP1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction