missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25518-1-AP-150UL
This item is not returnable.
View return policy
Description
Phosphoglycerate mutase (PGAM) catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). PGAM2 encodes muscle-specific PGAM subunit. Mutations in this gene cause muscle phosphoglycerate mutase eficiency, also known as glycogen storage disease X.Specifications
| PGAM2 | |
| Polyclonal | |
| Unconjugated | |
| PGAM2 | |
| BPG dependent PGAM 2, PGAM M, PGAM2, PGAMM, Phosphoglycerate mutase 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5224, 56012 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70250, P15259 | |
| PGAM2 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction