missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGAM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598051
This item is not returnable.
View return policy
Description
PGAM2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Phosphoglycerate mutase (PGAM) catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). PGAM2 encodes muscle-specific PGAM subunit. Mutations in this gene cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X.
Specifications
| PGAM2 | |
| Polyclonal | |
| Unconjugated | |
| Pgam2 | |
| BPG-dependent PGAM 2; D14Mgh1; EGK_13739; GSD10; Muscle-specific phosphoglycerate mutase; Pgam2; PGAMM; PGAM-M; Pgmut; Phosphoglycerate mutase 2; phosphoglycerate mutase 2 (muscle); phosphoglycerate mutase isozyme M; phosphoglycerate mutase muscle-specific subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24959, 5224, 56012 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.09 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70250, P15259, P16290 | |
| Pgam2 | |
| Recombinant Human Phosphoglycerate mutase 2 protein (1-253AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction