missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM2 Mouse anti-Human, Mouse, Clone: 5A7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59410-100UL
This item is not returnable.
View return policy
Description
PGAM2 Monoclonal specifically detects PGAM2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PGAM2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1020, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5224 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5A7 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| PGAM2 | |
| Recombinant human PGAM2 (1-253aa) purified from E. coli | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction