missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | PGAM2 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686065
|
Novus Biologicals
NBP2-38889-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18104479
|
Novus Biologicals
NBP2-38889 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGAM2 Polyclonal specifically detects PGAM2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PGAM2 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Proteases & Other Enzymes, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
| PGAM2 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P15259 | |
| 5224 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PMDEKHPYYNSISKERRYAGLK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title