missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM2 Antibody (5A7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59410H
This item is not returnable.
View return policy
Description
PGAM2 Monoclonal antibody specifically detects PGAM2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| PGAM2 | |
| Monoclonal | |
| HRP | |
| BPG-dependent PGAM 2, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, Muscle-specific phosphoglycerate mutase, PGAM-MGSD10, PGAMMMGC88743, phosphoglycerate mutase 2, phosphoglycerate mutase 2 (muscle), Phosphoglycerate mutase isozyme M | |
| Recombinant human PGAM2 (1-253aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Proteases & Other Enzymes, Signal Transduction | |
| 5224 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 5A7 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction